3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine ≥97%

Supplier: AOBChem USA

Warning

62404-1G 62404-500MG 62404-5G 62404-25G
76917-634EA 768.1 USD
76917-634 76917-636 76917-638 77656-718
3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine ≥97%
3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Br₂N₃
Storage Temperature: Ambient
MDL Number: MFCD20923265
CAS Number: 1352395-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR