3-Fluoro-2-iodo-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61996-1G 61996-25G 61996-500MG 61996-5G
76917-622EA 167.63 USD
76917-622 76917-624 76917-626 76917-628
3-Fluoro-2-iodo-4-methoxybenzaldehyde ≥97%
3-Fluoro-2-iodo-4-methoxybenzaldehyde
Formula: C₈H₆FIO₂
MW: 280.0349 g/mol
Storage Temperature: Ambient
CAS Number: 2092608-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR