3,4-Dibromo-5-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61882-25G 61882-5G
76917-614EA 595.17 USD
76917-614 76917-616
3,4-Dibromo-5-methyl-1H-indazole ≥97%
3,4-Dibromo-5-methyl-1H-indazole
Formula: C₈H₆Br₂N₂
MW: 289.9546 g/mol
Storage Temperature: Ambient
CAS Number: 2733056-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR