3,4-Dibromo-7-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61878-1G 61878-25G 61878-500MG 61878-5G
76917-598EA 342.68 USD
76917-598 76917-600 76917-602 76917-604
3,4-Dibromo-7-chloro-1H-indazole ≥97%
3,4-Dibromo-7-chloro-1H-indazole
Formula: C₇H₃Br₂ClN₂
MW: 310.3731 g/mol
Storage Temperature: Refrigerator
CAS Number: 2385648-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR