3,5-Dibromo-6-fluoro-1H-indazole ≥97% research grade

Supplier: AOBChem USA

Warning

61875-1G 61875-500MG 61875-5G
76917-588EA 527.27 USD
76917-588 76917-590 76917-592
3,5-Dibromo-6-fluoro-1H-indazole ≥97% research grade
3,5-Dibromo-6-fluoro-1H-indazole
Formula: C₇H₃Br₂FN₂
Storage Temperature: Ambient
CAS Number: 1596649-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR