3,5-Dibromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61872-1G 61872-25G 61872-500MG 61872-5G
76917-572EA 124.13 USD
76917-572 76917-574 76917-576 76917-578
3,5-Dibromo-1H-indazole ≥97%
3,5-Dibromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00159919
CAS Number: 40598-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR