3,6-Dibromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61869-1G 61869-25G 61869-500MG 61869-5G
76917-556EA 144.29 USD
76917-556 76917-558 76917-560 76917-562
3,6-Dibromo-1H-indazole ≥97%
3,6-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
MW: 275.928 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781598
CAS Number: 885521-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR