3-Bromo-2,4,6-trifluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61103-1G 61103-25G 61103-500MG 61103-5G
76917-498EA 184.6 USD
76917-498 76917-500 76917-502 76917-504
3-Bromo-2,4,6-trifluorobenzaldehyde ≥97%
3-Bromo-2,4,6-trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₂BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD11845971
CAS Number: 1160573-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR