3-Bromo-5-chloro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

60785-1G 60785-25G 60785-500MG 60785-5G
76917-482EA 131.56 USD
76917-482 76917-484 76917-486 76917-488
3-Bromo-5-chloro-N-methoxy-N-methylbenzamide ≥97%
3-Bromo-5-chloro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD28384163
CAS Number: 1700202-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR