3-Fluoro-5-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

59992-25G 59992-5G
76917-458EA 492.26 USD
76917-458 76917-460
3-Fluoro-5-iodopyridine ≥97%
3-Fluoro-5-iodopyridine
Formula: C₅H₃FIN
Storage Temperature: Ambient
MDL Number: MFCD08457780
CAS Number: 757950-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR