3,4-Dibromothiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

59186-1G 59186-25G 59186-5G
76917-444EA 64.72 USD
76917-444 76917-446 76917-448
3,4-Dibromothiophene-2-carboxylic acid ≥97%
3,4-Dibromothiophene-2-carboxylic acid
Formula: C₅H₂Br₂O₂S
Storage Temperature: Refrigerator
CAS Number: 7311-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR