3-Formyl-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58212-1G 58212-500MG 58212-5G
76917-418EA 1019.53 USD
76917-418 76917-420 76917-422
3-Formyl-5-(trifluoromethoxy)benzoic acid ≥97%
3-Formyl-5-(trifluoromethoxy)benzoic acid
Formula: C₉H₅F₃O₄
Storage Temperature: Refrigerator
CAS Number: 2167993-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR