3-Fluoro-5-formyl-4-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

57939-1G 57939-500MG 57939-5G
76917-394EA 850.85 USD
76917-394 76917-396 76917-398
3-Fluoro-5-formyl-4-methoxybenzoic acid ≥95%
3-Fluoro-5-formyl-4-methoxybenzoic acid
Formula: C₉H₇FO₄
CAS Number: 2137718-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR