3-Bromo-N-methoxy-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

57865-1G 57865-25G 57865-500MG 57865-5G
76917-380EA 233.4 USD
76917-380 76917-382 76917-384 76917-386
3-Bromo-N-methoxy-5-nitrobenzamide ≥97%
3-Bromo-N-methoxy-5-nitrobenzamide
Formula: C₈H₇BrN₂O₄
Storage Temperature: Ambient
CAS Number: 1515621-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR