4,5-Difluoro-2-formylbenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

57858-1G 57858-500MG 57858-5G
76917-370EA 467.86 USD
76917-370 76917-372 76917-374
4,5-Difluoro-2-formylbenzoic acid ≥97% research grade
4,5-Difluoro-2-formylbenzoic acid
Formula: C₈H₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD23161461
CAS Number: 1374150-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR