4,6-Difluoroindole-2-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4,6-Difluoro-1H-indole-2-carboxylic acid
57429-25G
57429-5G
76917-338EA
328.88
USD
76917-338
76917-340
4,6-Difluoroindole-2-carboxylic acid ≥97%
4,6-Difluoroindole-2-carboxylic acid
Formula:
C₉H₅F₂NO₂ MW: 197.1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863158 CAS Number: 247564-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...