4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

57061-1G 57061-25G 57061-5G
76917-304EA 62.6 USD
76917-304 76917-306 76917-308
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde ≥97%
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde
Formula: C₅H₅BrN₂O
MW: 189.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03074341
CAS Number: 473528-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR