3-Chloro-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-m-cresol, 3-Chloro-5-hydroxytoluene

56998-25G 56998-5G
76917-292EA 289.63 USD
76917-292 76917-294
3-Chloro-5-methylphenol ≥97%
3-Chloro-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD08234650
CAS Number: 58291-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR