3-Bromo-5-(difluoromethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

56729-1G 56729-25G 56729-500MG 56729-5G
76917-268EA 272.66 USD
76917-268 76917-270 76917-272 76917-274
3-Bromo-5-(difluoromethoxy)pyridine ≥97%
3-Bromo-5-(difluoromethoxy)pyridine
Formula: C₆H₄BrF₂NO
MW: 224.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185846
CAS Number: 342602-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR