3-Ethoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

56263-1G 56263-25G 56263-500MG 56263-5G
76917-254EA 126.25 USD
76917-254 76917-256 76917-258 76917-260
3-Ethoxy-2-naphthoic acid ≥97%
3-Ethoxy-2-naphthoic acid
Formula: C₁₃H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD8261051
CAS Number: 54245-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR