4,6-Dihydroxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid

55147-25G 55147-5G
76917-188EA 488.02 USD
76917-188 76917-190
4,6-Dihydroxynicotinic acid ≥97%
4,6-Dihydroxynicotinic acid
Formula: C₆H₅NO₄
MW: 155.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234168
CAS Number: 5466-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR