3-Ethyl-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

54856-1G 54856-500MG 54856-5G
76917-174EA 541.06 USD
76917-174 76917-176 76917-178
3-Ethyl-5-methylbenzaldehyde ≥95%
3-Ethyl-5-methylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.202 g/mol
Storage Temperature: Ambient
CAS Number: 3132-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR