3-Formyl-4,5-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54711-1G 54711-25G 54711-500MG 54711-5G
76917-160EA 406.33 USD
76917-160 76917-162 76917-164 76917-166
3-Formyl-4,5-dimethoxybenzoic acid ≥97%
3-Formyl-4,5-dimethoxybenzoic acid
Formula: C₁₀H₁₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD24483034
CAS Number: 27203-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR