3-Iodo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

53984-1G 53984-25G 53984-500MG 53984-5G
76917-108EA 310.85 USD
76917-108 76917-110 76917-112 76917-114
3-Iodo-5-methylphenol ≥97%
3-Iodo-5-methylphenol
Formula: C₇H₇IO
Storage Temperature: Ambient
CAS Number: 1073339-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR