3-(6-Methoxypyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53500-1G 53500-25G 53500-500MG 53500-5G
76917-058EA 277.96 USD
76917-058 76917-060 76917-062 76917-064
3-(6-Methoxypyridin-3-yl)benzaldehyde ≥97%
3-(6-Methoxypyridin-3-yl)benzaldehyde
Formula: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD06801892
CAS Number: 893736-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR