3-Fluoro-4-formyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53456-1G 53456-25G 53456-500MG 53456-5G
76917-044EA 236.59 USD
76917-044 76917-046 76917-048 76917-050
3-Fluoro-4-formyl-2-methoxybenzoic acid ≥97%
3-Fluoro-4-formyl-2-methoxybenzoic acid
Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34564236
CAS Number: 2383245-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR