3-Ethyl-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53429-1G 53429-500MG 53429-5G
76917-030EA 1181.85 USD
76917-030 76917-032 76917-034
3-Ethyl-5-formylbenzoic acid ≥97%
3-Ethyl-5-formylbenzoic acid
Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20660433
CAS Number: 1187028-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR