4-(Ethylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53322-1G 53322-25G 53322-5G
76917-014EA 89.12 USD
76917-014 76917-016 76917-018
4-(Ethylthio)benzaldehyde ≥97%
4-(Ethylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀OS
MW: 166.24
Storage Temperature: Refrigerator
MDL Number: MFCD01318145
CAS Number: 84211-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR