3-Bromo-2-hydroxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53159-1G 53159-500MG 53159-5G
76917-000EA 631.24 USD
76917-000 76917-002 76917-004
3-Bromo-2-hydroxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-2-hydroxy-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD4039295
CAS Number: 886762-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR