3-Bromo-5-iodophenol ≥95%

Supplier: AOBChem USA

Warning

53112-1G 53112-25G 53112-500MG 53112-5G
76916-986EA 144.29 USD
76916-986 76916-988 76916-990 76916-992
3-Bromo-5-iodophenol ≥95%
3-Bromo-5-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MW: 298.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405397
CAS Number: 570391-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR