3,4-Dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

52733-1G 52733-25G 52733-5G
76916-912EA 106.09 USD
76916-912 76916-914 76916-916
3,4-Dimethoxy-1,1'-biphenyl ≥97%
3,4-Dimethoxy-1,1'-biphenyl
Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00185066
CAS Number: 17423-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR