4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Bromophenyl)benzaldehyde, 4'-Bromo-4-formylbiphenyl, 4'-Bromo-[1,1'-biphenyl]-4-carbaldehyde

52715-25G 52715-5G
76916-894EA 630.18 USD
76916-894 76916-896
4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde ≥97%
4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉BrO
MW: 261.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980925
CAS Number: 50670-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR