4,5-Dichloronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

52439-1G 52439-25G 52439-500MG 52439-5G
76916-868EA 263.11 USD
76916-868 76916-870 76916-872 76916-874
4,5-Dichloronicotinic acid ≥97%
4,5-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01861987
CAS Number: 473837-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR