3-(tert-Butyl)-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52385-1G 52385-25G 52385-5G
76916-858EA 96.55 USD
76916-858 76916-860 76916-862
3-(tert-Butyl)-4-methoxybenzoic acid ≥97%
3-(tert-Butyl)-4-methoxybenzoic acid
Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16307217
CAS Number: 66737-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR