3-Fluoro-2-(methoxymethoxy)-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52151-1G 52151-500MG 52151-5G
76916-844EA 471.04 USD
76916-844 76916-846 76916-848
3-Fluoro-2-(methoxymethoxy)-4-methylbenzaldehyde ≥97%
3-Fluoro-2-(methoxymethoxy)-4-methylbenzaldehyde
Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34178619
CAS Number: 2643367-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR