3-Fluoro-2,6-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52040-1G 52040-500MG 52040-5G
76916-832EA 589.87 USD
76916-832 76916-834 76916-836
3-Fluoro-2,6-dimethylbenzoic acid ≥97%
3-Fluoro-2,6-dimethylbenzoic acid
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20650086
CAS Number: 26829-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR