3-(Difluoromethoxy)-2-fluoro-pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Difluoromethoxy)-2-fluoropyridine

51806-25G 51806-5G
76916-810EA 297.06 USD
76916-810 76916-812
3-(Difluoromethoxy)-2-fluoro-pyridine ≥97%
3-(Difluoromethoxy)-2-fluoro-pyridine
Formula: C₆H₄F₃NO
MW: 163.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185836
CAS Number: 1214377-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR