3-Iodo-7-(trifluoromethyl)-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
50693-1G
50693-25G
50693-500MG
50693-5G
76916-768EA
266.29
USD
76916-768
76916-770
76916-772
76916-774
3-Iodo-7-(trifluoromethyl)-1H-indazole ≥97%
3-Iodo-7-(trifluoromethyl)-1H-indazole
Formula:
C₈H₄F₃IN₂ MW: 312.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26519160 CAS Number: 1892533-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...