3-Bromo-2-ethoxy-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

50176-1G 50176-25G 50176-500MG 50176-5G
76916-754EA 173.99 USD
76916-754 76916-756 76916-758 76916-760
3-Bromo-2-ethoxy-6-fluorobenzaldehyde ≥97%
3-Bromo-2-ethoxy-6-fluorobenzaldehyde
Formula: C₉H₈BrFO₂
MDL Number: MFCD30184473
CAS Number: 1780186-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR