3-Bromo-2,6-dichloro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

49470-1G 49470-25G 49470-500MG 49470-5G
76916-688EA 411.63 USD
76916-688 76916-690 76916-692 76916-694
3-Bromo-2,6-dichloro-4-methylpyridine ≥97%
3-Bromo-2,6-dichloro-4-methylpyridine
Formula: C₆H₄BrCl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD28902504
CAS Number: 1935951-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR