3-Bromo-2-chloro-5-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

49377-1G 49377-25G 49377-500MG 49377-5G
76916-674EA 233.4 USD
76916-674 76916-676 76916-678 76916-680
3-Bromo-2-chloro-5-fluorobenzaldehyde ≥97% research grade
3-Bromo-2-chloro-5-fluorobenzaldehyde
Formula: C₇H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28166247
CAS Number: 1807017-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR