4-Bromo-2-chloro-5-methylaniline ≥97%

Supplier: AOBChem USA

Warning

49198-1G 49198-25G 49198-500MG 49198-5G
76916-658EA 183.54 USD
76916-658 76916-660 76916-662 76916-664
4-Bromo-2-chloro-5-methylaniline ≥97%
4-Bromo-2-chloro-5-methylaniline
Formula: C₇H₇BrClN
MW: 220.494 g/mol
Storage Temperature: Ambient
CAS Number: 888485-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR