3-Bromo-2-fluoro-5-iodobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

48874-1G 48874-25G 48874-500MG 48874-5G
76916-632EA 284.33 USD
76916-632 76916-634 76916-636 76916-638
3-Bromo-2-fluoro-5-iodobenzoic acid ≥97% research grade
3-Bromo-2-fluoro-5-iodobenzoic acid
Formula: C₇H₃BrFIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31620891
CAS Number: 1992023-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR