4-Benzyloxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48768-25G 48768-5G
76916-622EA 254.62 USD
76916-622 76916-624
4-Benzyloxy-2-methoxybenzaldehyde ≥97%
4-Benzyloxy-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD01075699
CAS Number: 58026-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR