3-Chloro-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48636-1G 48636-25G 48636-5G
76916-610EA 84.88 USD
76916-610 76916-612 76916-614
3-Chloro-4-propoxybenzoic acid ≥97%
3-Chloro-4-propoxybenzoic acid
Formula: C₁₀H₁₁ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08691861
CAS Number: 76327-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR