3-Cyano-2-fluoro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

48356-1G 48356-25G 48356-500MG 48356-5G
76916-596EA 233.4 USD
76916-596 76916-598 76916-600 76916-602
3-Cyano-2-fluoro-4-methoxyphenylboronic acid ≥97%
3-Cyano-2-fluoro-4-methoxyphenylboronic acid
Formula: C₈H₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD33403879
CAS Number: 2573124-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR