4-Bromo-2,3-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48296-1G 48296-25G 48296-500MG 48296-5G
76916-580EA 274.78 USD
76916-580 76916-582 76916-584 76916-586
4-Bromo-2,3-dichlorobenzaldehyde ≥97%
4-Bromo-2,3-dichlorobenzaldehyde
Formula: C₇H₃BrCl₂O
MW: 253.908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686180
CAS Number: 1349715-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR