3-Bromo-N-methoxy-N-methylbenzamide ≥95%
Supplier: AOBChem USA
|
Warning
|
48104-25G
48104-5G
76916-574EA
310.85
USD
76916-574
76916-576
3-Bromo-N-methoxy-N-methylbenzamide ≥95%
3-Bromo-N-methoxy-N-methylbenzamide
Formula:
C₉H₁₀BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD02684302 CAS Number: 207681-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...