3-Bromo-6-chloro-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

47911-25G 47911-5G
76916-540EA 254.62 USD
76916-540 76916-542
3-Bromo-6-chloro-2-fluoropyridine ≥97%
3-Bromo-6-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD06798230
CAS Number: 885952-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR