3-(Benzyloxy)-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

47724-1G 47724-25G 47724-500MG 47724-5G
76916-524EA 170.81 USD
76916-524 76916-526 76916-528 76916-530
3-(Benzyloxy)-2-methylbenzoic acid ≥95%
3-(Benzyloxy)-2-methylbenzoic acid
Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD22041842
CAS Number: 208986-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR